Sample cups for analysers
Leverancier: PerkinElmer
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
B0510397 B3001566 B3001567 B0087600 B0119079
PERCB0510397EA 218 EUR
PERCB0510397 PERCB3001566 PERCB3001567 PERCB0087600 PERCB0119079
Sample cups for analysers
Monsternamepotjes Monsternamepotjes voor analysatoren
Disposable.
These sample cups are of heavy-duty construction to prevent cracking or tipping. Their conical design allows for use of nearly the entire sample. For concentrated acids including HF for analyses.