Retort rods, unthreaded
Leverancier: HARVARD APPARATUS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
53-2570-R 53-2512-RM 53-2530-R 53-2540-R 53-2550-R 53-2560-R 53-2580-R
241-2117EA 0 EUR
241-2117 241-2111 241-2113 241-2114 241-2115 241-2116 241-2118
Retort rods, unthreaded
Statiefstaven
Stainless steel.
  • Brilliantly polished stainless steel with good surface hardness that will not score from clamp binding screws
  • Hollow, lightweight rods (Ø 13 mm) with continuously welded walls (1,6 mm thick)
  • Good chemical resistance
  • Red caps