Bottles, wide neck, with screw cap
Leverancier: LAMAPLAST
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
35 37 36
LAMA35EA 6.6 EUR
LAMA35 LAMA37 LAMA36
Bottles, wide neck, with screw cap
Flessen Kunststof flessen
HDPE, translucent.
  • Equipped with inner-cap
  • Emptying guaranteed by the inclination of the shoulders
  • For food use
Suitable for the storage of dense liquids, granulates and powders. The wide mouth allows an easy filling.