Jerricans, narrow neck, HDPE, UN Approved
Leverancier: LAMAPLAST
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
4122 4121 4119 4118 4114 4113 4112 4111 4110 4115
216-2343EA 29.4 EUR
216-2343 216-2342 216-2341 216-2340 216-2338 216-2337 216-2336 216-2335 216-2334 216-2339
Jerricans, narrow neck, HDPE, UN Approved
Flessen Jerrycans
Stackable jerricans, made of HDPE.
  • Safe and perfect application of labels on the surface of walls
  • Concave bottom allows easy transfer
  • Provided with handles
Fitted with self-sealing cap with PE foam gasket which ensures the tightness. Ideal for shipments of dangerous products into the UN rules, and in accordance with the regulation for road transport.