dNTP sets (dA, dC, dG, dT) 100 mM solutions
Leverancier: G Bioscienes
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
786-460 786-465 786-468 786-466 786-467
786-460EA 475 EUR
786-460 786-465 786-468 786-466 786-467
dNTP sets (dA, dC, dG, dT) 100 mM solutions
Reagentia voor nucleïnezuren Nucleotiden
Set consists of 100 mM aqueous solutions of dATP, dCTP, dGTP and dTTP each supplied in a separate vial. Also available separately.
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c40d4428cdfd614482e4f23c062c16d9d8242da2&vid=c40d4428cdfd614482e4f23c062c16d9d8242da2&prid=web&clid=SAPDAM