Rat single tissue blot, single species
Leverancier: G Bioscienes
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
TB03 TB06 TB09 TB12 TB15 TB18 TB21 TB24 TB27
TB03EA 720 EUR
TB03 TB06 TB09 TB12 TB15 TB18 TB21 TB24 TB27
Rat single tissue blot, single species
Reagentia voor elektroforese Western Blotting Reagents
This blot contains two protein lanes, a protein marker lane and a lane with one GenLysate™ sample. After the proteins are resolved, they are transferred to a PVDF membrane using Western Transfer Buffer. The blots are ready to be blocked and probed with the antibodies of choice.
  • Ready to screen
  • Single tissue rat blots