Bottles, wide neck, square, with screw cap
Leverancier: BURKLE
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
0338-1000 0338-0250 0338-0500
BURK0338-1000EA 4 EUR
BURK0338-1000 BURK0338-0250 BURK0338-0500
Bottles, wide neck, square, with screw cap
Flessen Kunststof flessen
HDPE, translucent.
  • Shatterproof
  • Leakproof
  • Special labelling areas (for labels or manual lettering)
Cost effective and ideal for packing and storing of liquids, powder and granules.