TRITON™ X-114
Leverancier: G Bioscienes
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
786-515 786-516
786-515EA 192 EUR
786-515 786-516
TRITON™ X-114
TRITON™ X-114
Triton® X-114 is a non ionic detergent with a low cloud point (23 °C) making it suitable for protein solubilisation with phase-partitioning of hydrophilic proteins from amphiphilic proteins.
MDL Nummer: MFCD00128254
CAS nummer: 9036-19-5
UN: 3082
ADR: 9,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0b5a918f8e54e5f28bf388009a2246e8ea1c0457&vid=b77cb06d36ba875c922850efbff0789827b2589d&prid=web&clid=SAPDAM