PCR plates, 96-well, low profile, Hard-Shell®
Leverancier: BIO RAD
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
HSP-9601 HSP-9611 HSP-9621 HSP-9631 HSP-9635 HSP-9641 HSP-9645 HSP-9655 HSP-9661 HSP-9665 HSP-9666 HSP-9901 HSS-9601 HSP-9955 HSS-9665 HSS-9901 HSS-9641
BRADHSP-9601EA 0 EUR
BRADHSP-9601 BRADHSP-9611 BRADHSP-9621 BRADHSP-9631 BRADHSP-9635 BRADHSP-9641 BRADHSP-9645 BRADHSP-9655 BRADHSP-9661 BRADHSP-9665 BRADHSP-9666 BRADHSP-9901 BRADHSS-9601 BRADHSP-9955 BRADHSS-9665 BRADHSS-9901 BRADHSS-9641
PCR plates, 96-well, low profile, Hard-Shell®
Microplaten PCR-platen
Hard-Shell® PCR plates are designed to prevent robotic handling problems due to plate warping. The hard shell prevents distortion and shrinkage due to the exposure of the plates to the high temperatures of thermal cycling or heat sealing.
  • Skirts and decks are moulded from rigid thermostable polymer
  • Thin walled wells are moulded from virgin PP
  • Raised rims around each well allow tight sealing