Brain tissue region blots, single species
Leverancier: G Bioscienes
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
TB40 TB41 TB57 TB59
TB40EA 2640 EUR
TB40 TB41 TB57 TB59
Brain tissue region blots, single species
Reagentia voor elektroforese Western Blotting Reagents
G-Biosciences offers ready-to-screen, normal tissue, brain tissue region blots. The brain tissue specific regions are prepared by carefully dissecting out anatomically and functionally distinct regions of adult mouse, rat, human, and primate brains.
Premade blots are prepared with the appropriate GenLysate™ and 50 μg protein is loaded onto a 4 to 20% denaturing polyacrylamide gel, along with a prestained molecular weight marker. After the proteins are resolved, they are transferred to a PVDF membrane, using Western transfer buffer.