Kidney tissue region blot, single species
Leverancier: G Bioscienes
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
TB42
TB43
TB42EA
1190
EUR
TB42
TB43
Kidney tissue region blot, single species
Reagentia voor elektroforese
Western Blotting Reagents
These are ready to screen, normal tissue, kidney tissue region blots from mouse and rat. The kidney tissue specific regions are prepared by carefully dissecting out anatomically and functionally distinct regions of adult mouse and rat kidney.