Eye tissue region blot, single species
Leverancier: G Bioscienes
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
TB44 TB45
TB44EA 1190 EUR
TB44 TB45
Eye tissue region blot, single species
Reagentia voor elektroforese Western Blotting Reagents
These are ready to screen, normal tissue, eye tissue region blots from rat and rabbit. The eye tissue specific regions are prepared by carefully dissecting out anatomically and functionally distinct regions of adult rat and rabbit eyes.