Pump tubing, Tygon® MHSL 2001
Leverancier: Saint Gobain Life Sciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
07MHSL-04L 07MHSL-15L 07MHSL-19L 07MHSLM09
VERN07MHSL-04LEA 263 EUR
VERN07MHSL-04L VERN07MHSL-15L VERN07MHSL-19L VERN07MHSLM09
Pump tubing, Tygon® MHSL 2001
Slangen Transfer Tubing
Transparent, plasticiser-free tubing with superior pump-life.
  • Superior flex life in peristaltic pumps
  • Chemically resistant to a wide range of fluids
Especially designed for MEK and other aggressive solvents.
Temperature resistant from −73 to +57 °C
Hardness (Shore durometer A): 69
Density: 0,88
Certificaten: Meets FDA criteria for food contact