Clear inserts, polyester (PET) membrane, Transwell®
Leverancier: Corning
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
3450 3452 3460 3462 3470 3472 3464 3467
734-1577EA 232.03 EUR
734-1577 734-1578 734-1579 734-1580 734-1581 734-1583 734-2607 CORN3467
Clear inserts, polyester (PET) membrane, Transwell®
Microplaten Insertplaten en inserts
Permeable supports, also known as cell culture inserts, are an essential tool for the study of both anchorage-dependent and independent cell lines.
  • w Produce a cell culture environment that closely resembles an in vivo state
  • Co-culture cells with or without cell-to-cell contact
  • Design a diversity of experiments using various pore sizes, membrane types, and coatings 10 µm transparent membrane
  • Treated for optimal cell attachment
  • Excellent visibility under phase contrast microscopy
  • Supplied in multiple well plates
  • Sterile
Allow polarised cells to carryMore Product Information