Manifold, Optimizer® ST2
Leverancier: Entegris
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Optimizer®
ENTEAMVXL42KTEA 0 EUR
ENTEAMVXL42KT ENTEAMVXL32KT ENTEAMVXL22KT ENTEAMVXL42ETT ENTEAMVXL22ET ENTEAMVXL32ET ENTEAMVXL42ET
Manifold, Optimizer® ST2
Spruitstukken
Stainless steel/PFA manifold for use with Optimizer® ST DEV and DI photochemical filters.
  • Compact, stable design provides easy installation or retrofit into existing tools
  • Rapid filter changeout with minimal exposure to process fluids