1-(3-sulphopropyl)pyridinium hydroxide inner salt , ND SB 201
Leverancier: G Bioscienes
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen: NDSB 201, 3-(1-Pyridinio)propanesulphonate, 3-(Pyridin-1-ium-1-yl)propane-1-sulphonate
DG080 DG081
DG080EA 297 EUR
DG080 DG081
1-(3-sulphopropyl)pyridinium hydroxide inner salt , ND SB 201
1-(3-sulphopropyl)pyridinium hydroxide inner salt
NDSB 201 is a zwitterionic compound. Unlike zwitterionic detergents, the hydrophobic group in NDSB 201 is too short to form micelles, even at 1 M concentrations. NDSB 201 can be used for the purification of proteins and solubilisation of protein samples for 2D gel electrophoresis.
Formule: C₈H₁₁NO₃S
MW: 201.24 g/mol
MDL Nummer: MFCD00064468
CAS nummer: 15471-17-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7f1a5aff6b8b7016de531a1fbf03fd947d4dedd6&vid=7f1a5aff6b8b7016de531a1fbf03fd947d4dedd6&prid=web&clid=SAPDAM