Destain solutions
Leverancier: G Bioscienes
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
786-526
786-527
786-526EA
146
EUR
786-526
786-527
Destain solutions
Reagentia voor elektroforese
Reagentia voor kleurverwijdering
G-Biosciences' Destain I contains approximately 45% methanol and 9% glacial acetic acid. G-Biosciences' Destain II contains approximately 5% methanol and 7% acetic acid.