Cooling packs, Freez’Pack
Leverancier: CAMPING GAZ
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
21628 39040
CAMP21628EA 6.1 EUR
CAMP21628 CAMP39040
Cooling packs, Freez’Pack
Koelelementen
This cooling pack is ideal for keeping drinks, food or medicines cool for longer in a passive hard or soft cooler. Its components include water, carboxymethyllcellulose, acticide and blue colourant.
  • Food-safe (non-toxic refrigerant)
  • Dishwasher proof
  • Reusable