Wattenbollen
Leverancier: VWR
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
VWR®
734-2671EA 272 EUR
734-2671
Wattenbollen
Glaswol en katoen
Thorough machine processing provides consistent uniformity of shape and size for maximum efficacy in vial plugging. These high quality balls are very effective at controlling mite infestation.
  • Past in smalle vials
Opgelet: *De product(en) op deze pagina vallen buiten het toepassingsgebied van de EU-richtlijn 2019/904