Sulphhydryl immobilisation kit for proteins and peptides
Leverancier: G Bioscienes
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
786-805 786-804
786-805EA 1050 EUR
786-805 786-804
Sulphhydryl immobilisation kit for proteins and peptides
Eiwitzuivering Protein Purification Kits
This Sulphhydryl Immobilisation Kit is designed for the coupling of sulphhydryl containing peptides to a solid agarose support. The sulphhydryl coupling resin columns utilises iodoacetyl groups that specifically react with free sulphhydryls to form covalent, permanent thioether bonds.
  • Simple and efficient coupling
  • Reduced steric hindrance ensures greater binding
  • Generates five reusable, spin format affinity columns
  • Specific conjugation through free sulphhydryls