Solenoid operated valves, VV and VV C series
Leverancier: VACUUBRAND
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
20674090 20674091 20674094
VACU20674090EA 350 EUR
VACU20674090 181-1631 VACU20674094
Solenoid operated valves, VV and VV C series
Ventielen Vacuümventielen
These straight-through valves with electromagnetic (solenoid) drive are used for operating cycles with short intervals and it is often used as vacuum control valves. The straightforward design provides a valve with low leakage rate.
  • Easy to clean
  • High operating cycles
  • Long term tightness even after long operating time
  • Version C are made of materials with chemical resistance
  • Versatile connection alternatives via small flange or hose nozzle
The valve sealing of the VV-B 6C is made of a special fluoroelastomer with better form stability than common PTFE, much higher chemical resistance than FKM and excellent long-term leak tightness characteristics.