Laboratory forceps, blunt
Leverancier: HAMMACHER KARL
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
HWC134-12 HWC135-14 HWC136-16
232-2155EA 58.8 EUR
232-2155 232-1920 232-2152
Laboratory forceps, blunt
Pincetten
Wironit steel (18/12 stainless steel with 18% chrome and 10 to 12% nickel).
  • Resistant to corrosion
  • Anti-magnetic