Protein reducing agents
Leverancier: G Bioscienes
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
786-077 786-25PR
786-077EA 386 EUR
786-077 786-25PR
Protein reducing agents
Eiwitextractie Reagentia voor eiwitextractie
Protein-S-S-Reductant™ uses TCEP (Tris [2-carboxyethyl] phosphine), a popular alternative to β-mercaptoethanol and DTT (dithiothreitol). TCEP improves stability, increases effectiveness, and reduces proteins over a wider range of pH, including lower acidic pHs. Protein-S-S-Reductant™ completely reduces stable disulfide bonds in less than 5 minutes at room temperature and is compatible with the protein alkylation reactions. This ready to use solution is at a neutral pH and stabilised for long-term storage. Simply supplement Protein-S-S-Reductant™ in place of DTT or β-mercaptoethanol and boil the sample.
OneQuant™ DTT are single aliquots of DTT that eliminate the need for weighing; preventing loss of reagent and saving time. Add 90 μl water to a single tube to generate a 0,5 M DTT solution. Supplied with 40 individual tubes.
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0811a592356bade33db17ad2b2b8dddbcf31bd5f&vid=c5b1125c4115c00a2b2201cdfa7b8b117245dc3e&prid=web&clid=SAPDAM