Carbohydrate coupling resin
Leverancier: G Bioscienes
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
786-808 786-807
786-808EA 380 EUR
786-808 786-807
Carbohydrate coupling resin
Koppelings- en labelingssystemen voor eiwitten
Carbohydrate coupling resin is designed for the covalent immobilisation of carbohydrate containing molecules, including glycoproteins, to agarose beads. It is ideal for the coupling of polyclonal antibodies as it allows for the optimal orientation of the antibodies for affinity purification.
The Carbohydrate Immobilization Kit is supplied with carbohydrate coupling resin columns for the preparation of 2 ml affinity columns, a coupling catalyst, SpinOUT™ GT-600 Desalting Columns for removing oxidising reagents, the oxidizing reagent (sodium meta-periodate) and relevant buffers.