Dot blot system, Enhancer™
Leverancier: G-Biosciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
786-163EA 2450 EUR
786-163
Dot blot system, Enhancer™
Blotapparaten
Enhancer™ dot blot system allows for the rapid application of up to 384 protein or nucleic acid samples on to a 12×9 cm membrane for subsequent analysis. Samples are applied to a nitrocellulose or nylon membrane by capillary action, resulting in concentration of sample due to minimal diffusion.
  • Increase dot blot sensitivity by 10-fold
  • Apply up to 384 protein or nucleic acid samples
  • No cross-contamination risk
  • No expensive electronic equipment required