Microscale organic chemistry kit, Quickfit®
Leverancier: SciLabware
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
ML1 ML/FE10 ML/FE25 ML/FP10 ML/FP25 ML/FR10 ML/C1 ML/C2 ML/FR25 ML/MF23 ML/RA3 ML/SH4 ML/ST
BIBBML1EA 410 EUR
BIBBML1 212-0618 212-0619 212-0620 212-0621 212-0622 201-3043 201-3036 212-0623 201-3061 271-0001 201-3007 201-3011
Microscale organic chemistry kit, Quickfit®
Kits voor microchemische experimenten
Microscale shemistry kit that is designed for organic chemistry experiments using milligram quantities of reagents. The components allow the construction of many assemblies including reflux, distillation, moisture sensitive reactions and fractional distillation.
  • Comprises 12 items of glassware that all feature 14/10 joint sizes for compatibility- see spare parts list
  • Other components compatible with the kit ar listed below under accessories
  • All components utilise a combination of standard Quickfit® ground glass joints together with screwthread couplings, sealing 'O' rings and plastic screw caps
  • The screwthread connector system eliminates the requirement for grease, saves on space, using clamps and vastly reduces the danger of glassware becoming loose during reactions
Leveringsinformatie: Supplied in a sturdy Gratnells tray with protective foam insert. Also included in the kit is a bag of spare caps, 'O' rings, sealing discs.