femtoCHROMO™- HRP Substrate for chromogenic detection
Leverancier: G Bioscienes
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
786-384 786-385 786-386 786-387 786-388
786-384EA 413 EUR
786-384 786-385 786-386 786-387 786-388
femtoCHROMO™- HRP Substrate for chromogenic detection
Koppelings- en labelingssystemen voor eiwitten
A ready to use modified TMB (3, 3’, 5, 5’-Tetramethylbenzidine) substrate is used that generates a dark blue precipitate in the presence of horseradish peroxidase.
  • Detects >20 ng
  • High signal to background ratio and reproducibility
Leveringsinformatie: Supplied with an enhanced blocking agent, BLOT-QuickBlocker™, and a concentrated [10X] washing buffer, femtoTBST™ Buffer, to ensure low background staining. Optional HRP labelled goat α-mouse or rabbit antibodies are also supplied.