Modular cart with flat bed, AlphaMop™
Leverancier: Texwipe
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
TX7046 TX7054
141-1050EA 1770 EUR
141-1050 141-1052
Modular cart with flat bed, AlphaMop™
Moppen
Cleaning Cart:
For use with AlphaMop™ stainless steel and plastic rectangular buckets, this cart has a protective side to keep your buckets in place and capture spills. Approved for use in the food industry. Autoclave safe. Use cart extension (TX7046E) to create a three bucket system.
Stainless steel bucket:
This seamless, stainless steel bucket is designed for use in critical manufacturing. Made from 100% Type 304 stainless steel.
Autoclave safe
Interior volume marks allow for accurate dilution
Select the stainless steel cart TX7046 and optional cart extension (TX7046E) to make a convenientMore Product Information