Zoutzuur concentrate for dilution to 0.1 mol (0.1N), DILUT-IT® Dissolution Media Concentrate, J.T.Baker®
Leverancier: Avantor
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
Synoniemen: Hydrochloric acid solution, Muriatic acid
D011-00EA 32.1 EUR
D011-00 D011-02 D011-04
Zoutzuur concentrate for dilution to 0.1 mol (0.1N), DILUT-IT® Dissolution Media Concentrate, J.T.Baker®
Zoutzuur
DILUT-IT dissolution media concentrates are produced from raw materials in accordance with USP/EP/BP/JP guidelines under validated processes in Avantor Performance Materials' ISO certified manufacturing facilities
Formule: HCl
MW: 36.46 g/mol
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011324
CAS nummer: 7647-01-0
EINECS: 231-595-7
UN: 1789
ADR: 8 II
Order Now
Specification Test Results
For Laboratory,Research,or Manufacturing Use 0.098 - 0.102
STORE AT ROOM TEMPERATURE
Appearance Passes Test
Density (g/mL) at 25°C, For Information Only
Normality at 26x dilution 0.098 - 0.102
DILUT-IT dissolution media concentrates are produced from raw materials in accordance with USP/EP/BP/JP guidelines under validated processes in Avantor Performance Materials' ISO certified manufacturing facilities