Cleaning trolleys, Clino®, CR6 FP series, with flat wringer
Leverancier: PFENNIG REINIGUNGSTECHNIK
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
3550200 3550300BLUE 3550100 3550300
141-1043EA 1590 EUR
141-1043 PPSA3550300BLUE 141-1042 PPSA3550300
Cleaning trolleys, Clino®, CR6 FP series, with flat wringer
Transportwagens
Process reliability and efficiency in technical cleanrooms with double rolling bucket system with minimal space requirements. Made of stainless steel flat wringer with defined pressure point for uniform residual moisture and are suitable for cleanrooms.
  • With flat wringer Clino Edelstahl
  • ISO-conformity
  • Highly efficient cleaning
  • Assured contamination prevention
  • Ergonomic
Weight: 19.49 kg
Rest: 80 x 54 x 102 cm
Working: 80 x 54 x 102 cm
Verpakking: Packing unit: 1
Carton size: 106 x 61 x 78 cm
Carton weight: 19.49 kg