Heating thermostats, JUVO
Leverancier: JUCHHEIM Laborgerate GmbH
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
55020.11 55040.11
JUCH55020.11EA 8550 EUR
JUCH55020.11 JUCH55040.11
Heating thermostats, JUVO
Circulatiethermostaten Gesloten circulatiesystemen Verwarmingscirculatoren
These thermostats have a special design that ensures trouble-free continuous operation at temperatures up to 350 °C. They are built in stainless steel, with a glandless pump, a visual level gauge and a temperature limiter. If required, an expansion chamber containing a float switch can be fitted.
A double layer of heat transfer fluid in these thermostats suppress the oxidative decomposition of heat transfer fluids over 200 °C.
Marlotherm SH® Dowtherm® or low viscosity silicone oil can be used as heat transfer fluids.
These basic units are fitted with one or two 2 kW heating elements.
Certificaten: Complies with safety level 2 of DIN 12879.