Deutrium furnace kit, GTA 120
Leverancier: VARIAN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
9910028200 6310003700 190067900
VARI9910028200EA 172 EUR
VARI9910028200 VARI6310003700 VARI190067900
Deutrium furnace kit, GTA 120
Ovens Verassings- en moffelovens
GTA 120 deuterium graphite furnace AA operating supplies kit.
Leveringsinformatie: Includes graphite electrodes, 2 pairs, spare graphite shroud, Omega platform tubes, 5×10/pk, 100 µl syringe for programmable sampler dispenser PSD-120, 5 capillary assemblies for PSD 120, 5×10 ml plastic (LDPE) beakers for PSD, 2 ml sample vials, 2×1000/pk