Single wafer shippers, for 4" (100 mm) wafer handling, H 22/H 93 series
Leverancier: ENTEGRIS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
H9340-101-1116 H22-402-0615 H22-402A-0615 H22-401-62C02 H22-402-62C02 H22-401-0615 H22-40-0615_U H22-40-62C02
216-1589EA 0 EUR
216-1589 216-2346 216-2349 216-2345 216-2348 216-2344 ENTEH22-40-0615_U 216-2350
Single wafer shippers, for 4" (100 mm) wafer handling, H 22/H 93 series
Wafertransportsystemen Wafer verzendcontainers
H 22 Series
Conical pocket holds one wafer facedown contacting only the wafer’s edges.
  • Spring retains wafer and minimises particle generation
  • Stackable for efficient use of storage space
  • Loose or lock stacking, either tray on tray or cover on tray
  • Convenient screw on covers
  • Large area on cover provides space for labeling
H 93 Series
Holds one wafer face down contacting only the edge of the wafer. Contains four inserted cushions to retain wafer and minimise particle retention.
Tweezer slots on shipper bottom ease wafer removal
Stackable for efficient use of storage space
Large area on lid provides space for labelling
Pin-in socket hinge allows shipper to be opened completely without tipping backwards
PP replacement cushions available to retrofit existing packages
Clear PC material protects wafer and allows visual inspection without opening