Single wafer shipping boxes, for 150 mm wafer handling, H 22 series
Leverancier: ENTEGRIS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
H22-60-0615_U H22-602-0615 H22-601-0615_U H22-601-62C02 H22-60-62C02 H22-602-62C02
ENTEH22-60-0615_UEA 0 EUR
ENTEH22-60-0615_U 216-2360 216-2358 216-2359 216-2362 216-2361
Single wafer shipping boxes, for 150 mm wafer handling, H 22 series
Wafertransportsystemen Wafer verzendcontainers
For shipping and storage applications.
  • Holds one wafer face down in a conical pocket contacting only the edge of the wafer
  • Spring securely retains the wafer to minimise particle generation
  • Screw-on cover provides secure closure and easy opening
  • Large top surface provides a convenient space for labelling
  • Stackable shipping boxes allow efficient use of storage space