Mounting media (water-based)
Leverancier: IMMUNOLOGIC
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
4058 4059
4058.EA 65.3 EUR
4058. 4059.
Mounting media (water-based)
Kleurstoffen Microscopy and staining reagents
Imsol Mount is an aqueous mounting medium: Especially for immunohistochemistry slides stained with an alk. phos. method and other non solvent resistant chromogens.
  • Also suitable for immunofluorescence
  • Water based