VWR®, Microcentrifugebuisjes en schroefdoppen
Leverancier: VWR Collection
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
525-0647EA 55.5 EUR
525-0647 525-0649 525-0651 525-0643 525-0653 525-0654 525-0655 525-0656 525-0657 525-0658 525-0645 525-0659 525-0641
VWR®, Microcentrifugebuisjes en schroefdoppen
Buisjes Microcentrifugebuisjes
Made from high grade gamma resistance PP.
  • Autoclavable at 121 °C and freezable to –80 °C
  • Tubes can be centrifuged to 20000×g
  • DNase/RNase free and non pyrogenic
  • Caps accept colour coded inserts, and easily write on the frosted side of insert
  • O-rings made from silicone to insure a leakproof seal
  • 1,5 and 2,0 ml tubes provide easy and legible frosted area
  • Sustainable design based on virgin resins
Manufactured in a Class 100000 room environment.
Certificaten: ISO 13485:2003 en ISO 9001:2008.