UHPLC filter
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
27006 22180
555-6747EA 333 EUR
555-6747 555-7042
UHPLC filter
Filters Voorkolom filters en frits
Column protection for Hypersil GOLD™ 1,9 µm and Syncronis™ 1,7 µm columns without compromising performance.
  • Low volume filter cartridge design
  • Maintain peak shape
  • Minimal efficiency loss through dispersion