1-Propanol 99,0 GC, corrected for water content, BAKER ANALYZED®, J.T.Baker®
Leverancier: Avantor
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
Synoniemen: Propylic alcohol, Propan-1-ol, n-Propan-1-ol, Ethylcarbinol, n-Propyl alcohol, Propanol-1, n-Propanol, Propyl alcohol, 1-Propyl alcohol, 1-Hydroxypropane
9086-01EA 74.48 EUR
9086-01 9086-33
1-Propanol 99,0 GC, corrected for water content, BAKER ANALYZED®, J.T.Baker®
1-Propanol
Formule: H₃CCH₂CH₂OH
MW: 60.1 g/mol
Kookpunt: 97
Smeltpunt: –127 °C
Dichtheid: 0.8053 g/cm³ (20 °C)
Vlampunt: 15 (gesloten beker)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00002941
CAS nummer: 71-23-8
EINECS: 200-746-9
UN: 1274
ADR: 3 II
Merck Index: 13,07934
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ab459f809f7797b46eb6d275aa3b1e6e94dc70d1&vid=deb28b985024604344065eea93ff6c6a9dd5a431&prid=web&clid=SAPDAM
Order Now
Specification Test Results
For Laboratory,Research,or Manufacturing Use = 99.0 %
Assay (CH₃CH₂CH₂OH) (by GC, corrected for water) ≥99.0 %
Residue after Evaporation ≤0.0100 %