Spray gun
Leverancier: ENTEGRIS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
603-864 215-98 600037BL 221-02 600037 215-55 600037OR 500121 221-05
ENTE603-864EA 0 EUR
ENTE603-864 ENTE215-98 ENTE600037BL ENTE221-02 ENTE600037 ENTE215-55 ENTE600037OR ENTE500121 ENTE221-05
Spray gun
Spuitpistolen
High purity PFA and PTFE (all wetted parts).
  • Orifice: 4,8 mm
  • Connection: 1/4" or 1/2" FNPT
  • Pressure: max. 5516 mbar
Used in fume hoods and wet processing stations to help protect against corrosion.