Colour Reference Standards, RAL CLASSIC Colour Collections
Leverancier: Industrial Physics
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
VF6602
VF6605
VF6610
SIMEVF6602EA
26.7
EUR
SIMEVF6602
SIMEVF6605
SIMEVF6610
Colour Reference Standards, RAL CLASSIC Colour Collections
Standaarden
Standaarden voor kleurreferentie en spectroscopie
RAL 840-HR are the basic collection for matt colour shades and RAL 841-GL for glossy colour shades. Contains safety and signalling colours with the colour requirements of DIN regulations.