Gel drying Frames™
Leverancier: DIVERSIFIED BIOTECH
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
AB-110760 AB-110762
DIVEAB-110760EA 262 EUR
DIVEAB-110760 DIVEAB-110762
Gel drying Frames™
Geldrogers Geldrogers, drogen met lucht
Gel drying frames allow researchers for easy handling and crack free dry gels without the use of heat/vacuum dryers.
Leveringsinformatie: The complete kit includes 2 open frames, 2 solid back plates, 12 side clips and 50 sheets of pre-cut cellophane.