Coolers for Reagent Reservoirs
Leverancier: DIVERSIFIED BIOTECH
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
RESE-C025 RESE-C050 RESE-C100
DIVERESE-C025EA 88.4 EUR
DIVERESE-C025 DIVERESE-C050 DIVERESE-C100
Coolers for Reagent Reservoirs
Dozen Koelboxen en koeltassen
These benchtop coolers are placed under the reservoirs and keep reagents at ~5 °C for approximately 3 hours.
  • No need for a cumbersome ice bath, the frozen non toxic gel is contained in a sealed molded cavity
  • Extends safe working time for temperature sensitive reagents