NPP ELISA alkaline phosphatase substrate
Leverancier: Rockland ImmunochemicalsTotal Ratings: 0
Avg. Ratings: 0.0 out of 5
NPP ELISA alcaline phosphatase substrate is a chromogenic substrate used in immunosorbent assays.
Phosphatase-conjugated detection antibodies can act upon the pNPP to convert it to a yellow para-nitrophenol. This yellow product can be measured on a spectrophotometer at 405 nm.
Bestelinformatie: Diluent buffer NPP-B500 for NPP-10 has to be ordered separately.
Opgelet: Both the substrate and buffer have to be stored at 4 °C prior to opening and should be protected from moisture and light.