Immunoblotting kits, MaxTag™
Leverancier: Rockland Immunochemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
KIB004 KIB001 KIB002 KIB003 KID001 TW0020 TMBM-101
ROCKKIB004EA 265 EUR
ROCKKIB004 ROCKKIB001 ROCKKIB002 ROCKKIB003 ROCKKID001 ROCKTW0020 ROCKTMBM-101
Immunoblotting kits, MaxTag™
Reagentia voor elektroforese Western Blotting Reagents
These accurate and sensitive kits are ideal for Western blotting or dot blotting methods. They offer flexibility with two unique substrates: Bright Blue TMB substrate and Stable Brown DAB substrate. Components are colour-coded for error-free results. With MaxTag™ you can run up to 40 (10×10cm) immunoblots.
  • Rapid
  • Accurate
  • Flexible with two unique substrates
Leveringsinformatie: Complete kit includes ultra pure Tween® 20, IgG Bovine Serum Albumin, biotinylated anti-IgG antibody, enhanced Steptavadin-Peroxidase conjugate, TMB substrate and precipitating agent, DAB substrate tablets, hydrogen peroxide (H₂O₂) solution, reusable dropper bottles, control IgG, and instruction manual.