Accessories for refrigerated baths and circulators, Optima™
Leverancier: GRANT INSTRUMENTS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
VTP2-FLST12 VTP2-FLST26 VTP2-FLST5 VTP2-LT
GRANVTP2-FLST12EA 743 EUR
GRANVTP2-FLST12 GRANVTP2-FLST26 GRANVTP2-FLST5 462-0590
Accessories for refrigerated baths and circulators, Optima™
Circulatiethermostaten Open circulatiesystemen
High pressure pumps for use where applications require a higher pressure than the internal pump to maintain flow.
  • Low noise and compact design
Max. flow rate / max. pressure:
VTP1: 9 L/min, 1 bar
VTP2: 12 L/min, 1,66 bar
Pipe bore (inlet/outlet) 12,7 mm
Leveringsinformatie: Supplied with pipe connections and special lid for fitting to tank
Opgelet: Using these optional pumps will reduce working area and transfer additional heat to the bath, so the minimum temperature achievable (with or without) accessory cooling will be increased.
Order Now
Specificaties
Environmentally Preferable