Colorimetric test kit, sulphite, TESTOVAL®
Leverancier: Heyl GmbH & Co, Gebr.
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
410634
410635
HEYL410634EA
173
EUR
HEYL410634
HEYL410635
Colorimetric test kit, sulphite, TESTOVAL®
Milieutestkits
Visual colour test method using a comparator disc. Two reagents.