Magnetic stirring bar retrievers, PTFE Turbo
Leverancier: COWIE
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
004.2350.RE 004.2450.RE
COWI004.2350.REEA 60.4 EUR
COWI004.2350.RE COWI004.2450.RE
Magnetic stirring bar retrievers, PTFE Turbo
Roerders Roergereedschap
These retrivers are used for the removal of giant stirrer bars from vessels of all kinds.