Autoclave Bags, High Transparency
Leverancier: RATIOLAB
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
7001105 7001155 7001205 7001305
RATI7001105EA 242 EUR
RATI7001105 RATI7001155 RATI7001205 RATI7001305
Autoclave Bags, High Transparency
Zakken Afvalzakken
Bags of very strong PP-film with a broad seam at the bottom, for the disposal of contaminated labware.
  • Autoclavable at 121 °C (20 min.)
  • Film thickness 50 µm
For autoclaving, the bag must not be completely closed.
Opgelet: For autoclaving, the bag must not be closed firmly.
Order Now
Specificaties
Environmentally Preferable