ISE standards
Leverancier: Hach
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
30749 2406549 23249 2351749 2354153 2370853 1279249 2318149 1474949 2305442 2835153
HACH30749EA 42.1 EUR
HACH30749 HACH2406549 HACH23249 HACH2351749 HACH2354153 HACH2370853 HACH1279249 HACH2318149 HACH1474949 HACH2305442 HACH2835153
ISE standards
Standaarden Standaarden voor milieuanalyse
ISE standards are used to calibrate ion selective electrode measurement systems by coresponding the electrode output with the concentration of the analyte. Pairs of standards with concentrations that differ by a factor of ten are used for analysis.