Presence-Absence media in bottles
Leverancier: Hach
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
2401612 2323212 2401650 2323250
HACH2401612EA 85.2 EUR
HACH2401612 HACH2323212 HACH2401650 HACH2323250
Presence-Absence media in bottles
Media voor microbiologie Geprepareerde media voor microbiologie
Selective for Total Coliform Bacteria and Escherichia coli. Broth in pre-filled bottles.
  • Sterile
  • Disposable